Learn More
Exostosin-like 2/EXTL2 Rat anti-Mouse, Clone: 339509, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2536
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429515
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Exostosin-like 2/EXTL2 Monoclonal antibody MAB2536 Specifically detects Exostosin-like 2/EXTL2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Exostosin-like 2/EXTL2
339509
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
EXTL2
Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2 Asn43-Met330 Accession # Q9ES89
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2135
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9ES89
Alpha-1,4-N-acetylhexosaminyltransferase EXTL2, alpha-1,4-N-acteylhexosaminyltransferase, Alpha-GalNAcT EXTL2, EC 2.4.1.223, exostoses (multiple)-like 2, Exostosin like 2, exostosin-like 2, EXTL2, EXTR2, EXT-related protein 2, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, processed exostosin-like 2
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Exostosin-like 2/EXTL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse EXTL1, recombinant human (rh) Exostosin-like 2/EXTL2, or rhEXTL3 is observed.