missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPM2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534436
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is SW480 cell lysate. PA5-34436 can be used with blocking peptide PEP-1478.
EPM2A Polyclonal antibody specifically detects EPM2A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
EPM2A | |
Polyclonal | |
Unconjugated | |
EPM2A | |
epilepsy, progressive myoclonic epilepsy, type 2 gene alpha; epilepsy, progressive myoclonus type 2, Lafora disease (laforin); epilepsy, progressive myoclonus type 2A; epilepsy, progressive myoclonus type 2A, Lafora disease (laforin); EPM2; EPM2A; EPM2A glucan phosphatase, laforin; EPM2A, laforin glucan phosphatase; glucan phosphatase; Glycogen phosphatase; Lafora PTPase; laforin; LAFPTPase; MELF; RP1-28C20.2; Tg(TcraK,TcrbK)TG-BFlv; TG-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114005, 13853, 7957 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
B3EWF7, Q91XQ2, Q9WUA5 | |
EPM2A | |
A 17 amino acid synthetic peptide near the carboxy terminus of human EPM2A | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |