missing translation for 'onlineSavingsMsg'
Learn More

Epimorphin/Syntaxin 2 Rat anti-Human, Clone: 338824, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2936

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449795

  • £288.60 / 100µg

Estimated Shipment date
from Supplier 06-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Epimorphin/Syntaxin 2 Monoclonal antibody MAB2936 Specifically detects Epimorphin/Syntaxin 2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Epimorphin/Syntaxin 2
338824
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
STX2
E. coli-derived recombinant human Epimorphin/Syntaxin 2 Met1-Arg188 Accession # P32856
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2054
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P32856
EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Epimorphin is observed.
Human
Videos
Documents

For Research Use Only