missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA130293
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NCI-H460 cell lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EphA4 Polyclonal antibody specifically detects EphA4 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
EphA4 | |
Polyclonal | |
Unconjugated | |
EPHA4 | |
2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
13838, 2043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P54764, Q03137 | |
EPHA4 | |
KLH conjugated synthetic peptide between 875-904 amino acids from the C-terminal region of human EphA4 | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |