missing translation for 'onlineSavingsMsg'
Learn More

EphB6 Mouse anti-Human, Mouse, Clone: 465323, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3384

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440046

  • £288.60 / 100µg

Estimated Shipment
19-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EphB6 Monoclonal antibody MAB3384 Specifically detects EphB6 in Human, Mouse samples. It is validated for Western Blotting
Specifications

Specifications

EphB6
465323
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
EPHB6
Mouse myeloma cell line NS0-derived recombinant human EphB6 Val17-Thr579 Accession # O15197
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2051
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O15197
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse EphB6 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) EphB6 is observed, 25% cross-reactivity with recombinant human (rh) EphA10 is observed, and no cross-reactivity with rhEphA1, 2, 5, rhEphB3, 4, rmEphA1, 3, 4, 6, 7, 8, rmEphB2, 3, recombinant rat (rr) EphA5, or rrEphB1 is observed.
Human, Mouse
Videos
Documents

For Research Use Only