Learn More
EphA5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547735
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) EphA4, rmEphA3, rmEphA6, rmEphA7, rmEphA8, and recombinant rat EphB1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
EphA5 Polyclonal antibody specifically detects EphA5 in Human, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
EphA5 | |
Polyclonal | |
Unconjugated | |
EPHA5 | |
AI854630; ARCC2; AW125296; brain-specific kinase; BSK; CEK7; CEK-7; CTPA; CTPP1; ECK; eck-like sequence 1; EHK1; EHK-1; EK7; Ekh1; Els1; Els1.; epa5; EPH homology kinase 1; Eph receptor A5; EPHA5; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 5; EphrinA2; EphrinA5; HEK7; receptor protein-tyrosine kinase HEK7; Rek7; TYRO4; tyrosine-protein kinase receptor EHK-1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2044, 79208 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P54756, P54757 | |
EPHA5 | |
Mouse myeloma cell line NS0-derived recombinant rat EphA5 Met1-Gln573 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |