missing translation for 'onlineSavingsMsg'
Learn More

EphA4 Rat anti-Mouse, Clone: 107925, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB641

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441356

  • £337.93 / 500µg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EphA4 Monoclonal antibody MAB641 Specifically detects EphA4 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

EphA4
107925
Western Blot 1 ug/mL
Q03137
Cek8, EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, Sek1, Tyro1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse EphA4 in direct ELISAs and Western blots. Shows less than 5% cross-reactivity with recombinant mouse (rm) EphA3 and recombinant rat (rr) EphB1 and no cross-reactivity with recombinant human EphA1, rmEphA2, -A6, -A7, -A8, -B2, -B3, -B4, -B6, or rrEphA5.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
EPHA4
Mouse myeloma cell line NS0-derived recombinant mouse EphA4 Val20-Thr547 Accession # Q03137
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2043
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only