Learn More
Ephrin A2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547806
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-A1, recombinant human (rh) Ephrin-A3, rmEphrin-A4, rhEphrin-A5, rmEphrin-B1, rmEphrin-B2, and rhEphrin-B3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Ephrin A2 Polyclonal antibody specifically detects Ephrin A2 in Mouse samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Specifications
Ephrin A2 | |
Polyclonal | |
Unconjugated | |
EFNA2 | |
CEK7L; CEK7-L; CEK7-ligand; EFNA2; Elf1; ELF-1; EPH related receptor tyrosine kinase ligand 6; eph-related receptor tyrosine kinase ligand 6; ephrin A2; ephrin A-2; ephrin A6; EphrinA2; ephrin-A2; Epl6; EPLG6; HEK7 ligand; HEK7-L; Lerk6; LERK-6 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13637 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P52801 | |
EFNA2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A2 Met1-Asn184 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |