missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514574
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with non-human primate based on sequence homology.
EphA2 Polyclonal antibody specifically detects EphA2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EphA2 | |
Polyclonal | |
Unconjugated | |
EPHA2 | |
ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
13836, 1969 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P29317, Q03145 | |
EPHA2 | |
KLH conjugated synthetic peptide between 30-60 amino acids from the N-terminal region of human EphA2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |