missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA131986
Code : LE
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is mouse heart tissue.
EphB2 Polyclonal antibody specifically detects EphB2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
EphB2 | |
Polyclonal | |
Unconjugated | |
EPHB2 | |
CAPB; cEK5; chicken embryo kinase 5; chicken embryo kinase 5 protein; developmentally-regulated Eph-related tyrosine kinase; DRT; EFL6; EFNB3; EK5; ELK; ELK-related tyrosine kinase; embryo kinase 5 protein CEK5; EPH B2; EPH receptor B2; eph tyrosine kinase 3; EPHB2; EphB2/CTF1; EphB2/CTF2; EPHB3; EPH-like kinase 5; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPHT3; EPLG8; EPTH3; ERK; ETECK; ETK2; HEK2; Hek5; HEK-6; LERK8; MGC87492; NET; Neural kinase; Nuk; Nuk receptor tyrosine kinase; PCBC; Prkm5; protein-tyrosine kinase HEK5; Qek5; Renal carcinoma antigen NY-REN-47; RGD1564232; Sek3; TYRO5; TYRO6; Tyrosine-protein kinase receptor EPH-3; tyrosine-protein kinase receptor SEK-3; Tyrosine-protein kinase TYRO5 | |
Rabbit | |
Protein G | |
RUO | |
2048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.25-0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P29323 | |
EPHB2 | |
KLH conjugated synthetic peptide between 103-133 amino acids from the N-terminal region of human EphB2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |