missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547277
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse EPCR is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
EPCR Polyclonal antibody specifically detects EPCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EPCR | |
Polyclonal | |
Unconjugated | |
Procr | |
Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10544 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9UNN8 | |
Procr | |
Mouse myeloma cell line NS0-derived recombinant human EPCR Ser18-Ser210 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |