Learn More
EPB41L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529485
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4. Predicted reactivity: Mouse (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EPB41L2 Polyclonal antibody specifically detects EPB41L2 in Human samples. It is validated for Western Blot.Specifications
EPB41L2 | |
Polyclonal | |
Unconjugated | |
EPB41L2 | |
4.1G; 4.1-G; AW555191; band 4.1-like protein 2; D10Ertd398e; DKFZp781D1972; DKFZp781H1755; Epb4.1l2; Epb41l2; erythrocyte membrane protein band 4.1 like 2; erythrocyte membrane protein band 4.1 like-protein 2; erythrocyte membrane protein band 4.1-like 2; erythrocyte membrane protein band 4.1-like 2 retinal; erythrocyte protein band 4.1 like 2; erythrocyte protein band 4.1l2 long; erythrocyte protein band 4.1l2 middle; erythrocyte protein band 4.1-like 2; generally expressed protein 4.1; NBL2; OTTHUMP00000040262; RGD1563977; RP3-324N14.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.62 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O43491 | |
EPB41L2 | |
Recombinant fragment corresponding to a region within amino acids 107 and 384 of Human EPB41L2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |