missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41L2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518830
Code : A0
Additional Details : Weight : 0.01000kg
Description
EPB41L2 Polyclonal antibody specifically detects EPB41L2 in Human samples. It is validated for Immunohistochemistry.Specifications
EPB41L2 | |
Polyclonal | |
Unconjugated | |
EPB41L2 | |
4.1G; 4.1-G; AW555191; band 4.1-like protein 2; D10Ertd398e; DKFZp781D1972; DKFZp781H1755; Epb4.1l2; Epb41l2; erythrocyte membrane protein band 4.1 like 2; erythrocyte membrane protein band 4.1 like-protein 2; erythrocyte membrane protein band 4.1-like 2; erythrocyte membrane protein band 4.1-like 2 retinal; erythrocyte protein band 4.1 like 2; erythrocyte protein band 4.1l2 long; erythrocyte protein band 4.1l2 middle; erythrocyte protein band 4.1-like 2; generally expressed protein 4.1; NBL2; OTTHUMP00000040262; RGD1563977; RP3-324N14.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2037 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43491 | |
EPB41L2 | |
Synthetic peptide sequence (DYDPEEHGSID) corresponding to the internal amino acids of EPB41L2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |