missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518476
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
PTGER4 Polyclonal antibody specifically detects PTGER4 in Human samples. It is validated for Western Blot.Specifications
PTGER4 | |
Polyclonal | |
Unconjugated | |
PTGER4 | |
EP1; EP4; EP4R; MGC126583; PGE receptor EP4 subtype; PGE receptor, EP4 subtype; PGE2 receptor EP4 subtype; prostaglandin E receptor 4; prostaglandin E receptor 4 (subtype EP4); prostaglandin E2 receptor EP4 subtype; prostaglandin E2 receptor type 4; prostanoid EP4 receptor; Ptger; PTGER2; Ptger4; Ptgerep4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5734 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P35408 | |
PTGER4 | |
Synthetic peptide sequence (LEREVSKNPDLQA) corresponding to the internal amino acids of PTGER4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |