Learn More
p300 Monoclonal Antibody (7D8A6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515444
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15444 targets EP300 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15444 immunogen is purified recombinant fragment of EP300 expressed in E. Coli.
p300 Monoclonal antibody specifically detects p300 in Human samples. It is validated for ELISA, Western Blot.Specifications
p300 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q09472 | |
EP300 | |
Purified recombinant fragment of EP300 expressed in E. Coli. | |
RUO | |
2033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
7D8A6 | |
Unconjugated | |
EP300 | |
A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |