Learn More
ENPP-5 Mouse anti-Human, Clone: 500915, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5979
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13461166
  • £249.49 / 100µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
ENPP-5 Monoclonal antibody MAB5979 Specifically detects ENPP-5 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
ENPP-5
500915
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ENPP5
Mouse myeloma cell line NS0-derived recombinant human ENPP-5 Pro25-Ser430 Accession # Q9UJA9
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
59084
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q9UJA9
EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, ENPP5, KIAA0879, NPP5, NPP-5
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human ENPP-5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ENPP-2, rhENPP-7, or recombinant mouse ENPP-7 is observed.