Learn More
Engrailed-2 Mouse anti-Human, Clone: 279122, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2600
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459565
  • £261.37 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Engrailed-2 Monoclonal antibody MAB2600 Specifically detects Engrailed-2 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
Engrailed-2
Unconjugated
Murine
IgG2b
100μg
Primary
Human
279122
Purified
E.Coli-derived recombinant human Engrailed-2. Ser118-Glu333
Protein A or G purified from hybridoma culture supernatant