Learn More
Endorepellin/Perlecan Mouse anti-Human, Clone: 268908, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2364
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449385
  • £277.83 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Endorepellin/Perlecan Monoclonal antibody MAB2364 Specifically detects Endorepellin/Perlecan in Human samples. It is validated for Western Blotting
Specifications
Specifications
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan
268908
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
HSPG2
Mouse myeloma cell line NS0-derived recombinant human Endorepellin/Perlecan Arg3684-Ser4391 (Ser4003Thr) Accession # P98160
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3339
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P98160
basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, HSPG2, Perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Endorepellin/Perlecan in direct ELISAs and Western blots.