missing translation for 'onlineSavingsMsg'
Learn More

EphB1, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen PA112410

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11545962

  • £257.00 / 250µg


Estimated Shipment
27-06-2018
Add to basket

Description

Description

A suggested positive control for this product is human brain lysate.
Specifications

Specifications

ELK Rec. Tyrosine Kinase
Polyclonal
PBS with 0.08% sodium azide
P54762
ELK, EPHT2, FLJ37986, Hek6, NET
Sheep
IgG
250μg
Antibody
Polyclonal
Human
Immunoprecipitation
Unconjugated
EPHB1
Purified
EPHB1
Recombinant protein corresponding to residues 883-984 of C terminus of human ephB1.
Ammonium sulfate precipitation
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
2047
Videos