missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR1 Monoclonal Antibody (T.126.1), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA515009
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
EGR1 Monoclonal antibody specifically detects EGR1 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
EGR1 | |
Monoclonal | |
117 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P08046, P08154, P18146 | |
EGR1 | |
Synthetic peptide corresponding to the amino terminus of the sequence of human EGR1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
T.126.1 | |
Unconjugated | |
EGR1 | |
A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13653, 1958, 24330 | |
-20°C | |
Liquid |