Learn More
PROK1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547252
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, 100% cross reactivity with recombinant human EG-VEGF and recombinant mouse EG-VEGF is observed Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
PROK1 Polyclonal antibody specifically detects PROK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PROK1 | |
Polyclonal | |
Unconjugated | |
PROK1 | |
black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
192205, 246691, 84432 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P58294, Q14A28, Q8R414 | |
PROK1 | |
Mouse myeloma cell line NS0-derived recombinant rat EG-VEGF/PK1 Ala19-Phe105 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |