missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PROK1 Monoclonal Antibody (188608), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523785
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
PROK1 Monoclonal antibody specifically detects PROK1 in Human samples. It is validated for ELISA.Specifications
PROK1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P58294 | |
PROK1 | |
Mouse myeloma cell line NS0-derived recombinant human EG-VEGF/PK1 Ala20-Phe105 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
188608 | |
Unconjugated | |
PROK1 | |
black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186 | |
Mouse | |
Protein A/G | |
RUO | |
84432 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |