Learn More
EFTUD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530310
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain. Predicted reactivity: Mouse (100%), Zebrafish (97%), Xenopus laevis (98%), Chicken (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EFTUD2 Polyclonal antibody specifically detects EFTUD2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
EFTUD2 | |
Polyclonal | |
Unconjugated | |
EFTUD2 | |
116 kDa U5 small nuclear ribonucleoprotein component; 116kDa; Eftud2; elongation factor Tu GTP binding domain containing 2; elongation factor Tu GTP-binding domain-containing protein 2; hSNU114; KIAA0031; MFDGA; MFDM; SNRNP116; Snrp116; Snu114; SNU114 homolog; U5 small nuclear ribonucleoprotein 116 kDa; U5 snRNP-specific protein, 116 kDa; U5-116 kDa; U5-116KD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20624, 9343 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O08810, Q15029 | |
EFTUD2 | |
Recombinant fragment corresponding to a region within amino acids 633 and 933 of Human EFTUD2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |