Learn More
EEF1D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527666
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (91%), Rat (92%), Sheep (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EEF1D Polyclonal antibody specifically detects EEF1D in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
EEF1D | |
Polyclonal | |
Unconjugated | |
EEF1D | |
antigen NY-CO-4; EEF 1D; Eef1d; EF 1 delta; EF 1D; EF1D; EF-1D; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); FP1047; guanine nucleotide exchange protein; translation elongation factor 1-delta subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1936, 66656 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.91 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P29692, P57776 | |
EEF1D | |
Recombinant fragment corresponding to a region within amino acids 1 and 281 of Human EEF1D | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |