missing translation for 'onlineSavingsMsg'
Learn More

EDG2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA112306

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11515932

  • £430.00 / 100µg


Estimated Shipment
27-06-2018
Add to basket

Specifications

Specifications

EDG2
Polyclonal
PBS with 0.08% sodium azide
Q92633
LPA1, EDG2, VZG-1, LPA receptor 1, LPA-1
Rabbit
IgG
100μg
Antibody
Polyclonal
Human
Western Blot
Unconjugated
LPAR1
Purified
LPAR1
Synthetic peptide corresponding to the C-terminal of EDG2 receptor.
Ammonium sulfate precipitation
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
1902
Videos