missing translation for 'onlineSavingsMsg'
Learn More

EDAR Rat anti-Mouse, Clone: 132106, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB7451

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13451776

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EDAR Monoclonal antibody MAB7451 Specifically detects EDAR in Mouse samples. It is validated for ELISA Capture
Specifications

Specifications

EDAR
132106
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9R187
Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse EDAR in ELISAs. In sandwich immunoassays, less than 10% cross-reactivity with recombinant human EDAR is observed and no cross-reactivity with recombinant mouse (rm) EDA, rmTROY, or rhXEDAR is observed.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
EDAR
Mouse myeloma cell line NS0-derived recombinant mouse EDAR Glu27-Ile189 Accession # Q9R187
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10913
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only