missing translation for 'onlineSavingsMsg'
Learn More

EDA/Ectodysplasin Mouse anti-Human, Clone: 174806, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB9222

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461996

  • £337.93 / 500µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

EDA/Ectodysplasin Monoclonal antibody MAB9222 Specifically detects EDA/Ectodysplasin in Human samples. It is validated for ELISA Capture
Specifications

Specifications

EDA/Ectodysplasin
174806
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
NP_001005609
ectodysplasin A, ED1, EDAA1, EDAA2, Tabby, XHED, XLHED
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human EDA/Ectodysplasin (EDA-A1 and EDA-A2) in ELISAs. In sandwich immunoassays, approximately 50% cross-reactivity with recombinant mouse EDA is observed, and no cross-reactivity with recombinant human (rh) TNF-alpha , rhXEDAR, or rhEDAR is observed.
Human
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
EDA
Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1896
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only