missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECSCR Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA538003
Code : NEW
Additional Details : Weight : 0.00900kg
Description
ECSCR Polyclonal antibody specifically detects ECSCR in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ECSCR | |
Polyclonal | |
Unconjugated | |
ECSCR | |
1110006O17Rik; apoptosis regulator through modulating IAP expression; ARIA; Ecscr; ECSM2; endothelial cell surface expressed chemotaxis and apoptosis regulator; endothelial cell-specific chemotaxis regulator; Endothelial cell-specific molecule 2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
641700 | |
-20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q19T08 | |
ECSCR | |
Peptide with sequence C-NNGKQSLSAEKVL, from the C-terminus of ECSM2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |