Learn More
ECHDC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543230
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: FVQRLRGLMN DIASSAVMGL IETTRGLLPG AGGTQRLPRC LGVALAKELI Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
ECHDC2 Polyclonal antibody specifically detects ECHDC2 in Human samples. It is validated for Western Blot.Specifications
ECHDC2 | |
Polyclonal | |
Unconjugated | |
Echdc2 | |
1300017C12Rik; 2610009M20Rik; D4Ertd765e; ECHDC2; enoyl CoA hydratase domain containing 2; enoyl Coenzyme A hydratase domain containing 2; enoyl-CoA hydratase domain containing 2; enoyl-CoA hydratase domain-containing protein 2, mitochondrial; hypothetical protein LOC513795; id:ibd1032; RGD1308525; Unknown (protein for MGC:85763); wu:fc83b02; zgc:56321; zgc:85763 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55268 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86YB7 | |
Echdc2 | |
Synthetic peptide directed towards the middle region of human ECHDC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |