missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523297
Code : LD
Additional Details : Weight : 0.01000kg
Description
EBI3 Polyclonal antibody specifically detects EBI3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
EBI3 | |
Polyclonal | |
Unconjugated | |
EBI3 | |
cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; IL-27; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; IL35 subunit; interleukin-27 subunit beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10148, 50498, 680609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.05% sodium azide | |
O35228, Q14213 | |
EBI3 | |
A portion of amino acids 180-228 of mouse EBI3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |