missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR183 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523461
Code : LD
Additional Details : Weight : 0.01000kg
Description
GPR183 Polyclonal antibody specifically detects GPR183 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GPR183 | |
Polyclonal | |
Unconjugated | |
GPR183 | |
EBI 2; Ebi2; EBV-induced gene 2; EBV-induced G-protein coupled receptor 2; EBV-induced G-protein coupled receptor 2 homolog; Epstein-Barr virus induced gene 2; Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor); epstein-Barr virus-induced G-protein coupled receptor 2; Epstein-Barr virus-induced G-protein coupled receptor 2 homolog; G protein-coupled receptor 183; GPR183; G-protein coupled receptor 183; RP11-461N23.2 | |
Rabbit | |
Protein A | |
RUO | |
1880 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P32249 | |
GPR183 | |
A portion of amino acids 300-350 of human EBI2 | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |