missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV Type 16 E7 Monoclonal Antibody (3D6F1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515439
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15439 targets E7 in indirect ELISA, WB applications and shows reactivity with Tag samples. The MA5-15439 immunogen is purified recombinant fragment of E7 expressed in E. Coli.<br>.
HPV Type 16 E7 Monoclonal antibody specifically detects HPV Type 16 E7 in Virus samples. It is validated for ELISA, Western Blot.Specifications
HPV Type 16 E7 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Mouse | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
3D6F1 | |
Unconjugated | |
HPV 16-E7; HPV16E7; human papillomavirus | |
Purified recombinant full-lenght E7 expressed in E. Coli. | |
RUO | |
Virus | |
Antibody | |
IgG1 |