Learn More
E2F1 Monoclonal Antibody (KH95), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514344
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-14344 targets E2F-1 Transcription Factor in gel shift, immunofluorescence, immunohistochemistry (paraffin), immunoprecipitation, and Western blot applications and shows reactivity with Human and mouse samples. The MA5-14344 immunogen is recombinant human E2F-1 protein.
E2F1 Monoclonal antibody specifically detects E2F1 in Human, Mouse samples. It is validated for Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
E2F1 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q01094, Q61501 | |
E2F1 | |
Recombinant human E2F-1 protein | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
KH95 | |
Unconjugated | |
E2F1 | |
E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1 | |
Mouse | |
Protein A | |
RUO | |
13555, 1869 | |
4° C | |
Liquid |