Learn More
E. coli serotype O/K Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA17213
Code : LD
Additional Details : Weight : 1.24740kg
Description
PA1-7213 detects all "e;O"e; and "e;K"e; antigenic serotypes of Escherichia coli. This antibody is unabsorbed and will cross react with related enterobacteriaceae. PA1-7213 is suggested for use in ELISA procedures. The PA1-7213 immunogen is mixtures of all antigenic serotypes. Recommended short term (6 months) storage is liquid at 2-8 oC. For longer term storage, aliquot and freeze.
E. coli serotype O/K Polyclonal antibody specifically detects E. coli serotype O/K in Bacteria samples. It is validated for ELISA.Specifications
E. coli serotype O/K | |
Polyclonal | |
Unconjugated | |
Escherichia coli | |
Mixtures of all antigenic serotypes. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |