missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. coli serotype O157 Monoclonal Antibody (L65G), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA518198
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Purity is >90%. Recommended pairs for ELISA: MA5-18198 (capture) and MA5-18196 (conjugated, detection).
E. coli serotype O157 Monoclonal antibody specifically detects E. coli serotype O157 in Bacteria samples. It is validated for ELISA, Immunocytochemistry.Specifications
E. coli serotype O157 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
L65G | |
Unconjugated | |
Escherichia coli | |
E. coli native protein | |
100 μg | |
Primary | |
4° C | |
Liquid |