Learn More
E. coli serotype O Monoclonal Antibody (HEC10-165.3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110695
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10695 detects E. coli from E. coli O2a/2b, O7, O11, O18, O44, O75, O112, and O125 samples. MA1-10695 has been successfully used in ELISA applications. The MA1-10695 immunogen is E. coli serotypes: O18, O44, O112, O125.
E. coli serotype O Monoclonal antibody specifically detects E. coli serotype O in Bacteria samples. It is validated for ELISA.Specifications
E. coli serotype O | |
Monoclonal | |
0.8 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA | |
HEC10-165.3 | |
Unconjugated | |
Escherichia coli | |
E. coli serotypes: O18, O44, O112, O125. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |