missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. coli J5 LPS Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA185648
Code : LD
Additional Details : Weight : 0.01000kg
Description
Sheep anti Escherichia coli antibody recognizes the J5 mutant of E.
E. coli J5 LPS Polyclonal antibody specifically detects E. coli J5 LPS in Bacteria samples. It is validated for ELISA.Specifications
E. coli J5 LPS | |
Polyclonal | |
Unconjugated | |
Escherichia coli | |
Heat inactivated Escherichia coli J5 (Rc mutant; ATCC). | |
RUO | |
Bacteria | |
Antibody | |
IgG |
ELISA | |
Conc. Not Determined | |
whole serum with <0.1% sodium azide | |
Sheep | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |