Learn More
E. coli serotype O/K Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA173030
Code : LD
Additional Details : Weight : 0.01000kg
Description
The PA1-73030 antibody reacts with E. coli. And crossreacts with related enterobacteriaceae. PA1-73030 has been successfully used in ELISA applications. The PA1-73030 antibody was raised against O and K antigenic serotypes (All types) of Escherichia coli. Recommended short term storage of less than six months, keep at 2-8oC. For long term storage, store at -20oC.
E. coli serotype O/K Polyclonal antibody specifically detects E. coli serotype O/K in Bacteria samples. It is validated for ELISA.Specifications
E. coli serotype O/K | |
Polyclonal | |
HRP | |
Escherichia coli | |
O and K antigenic serotypes (All types) of Escherichia coli. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA | |
1-2 mg/mL | |
PBS with 10mg/mL BSA and 0.002% thimerosal | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |