Learn More
E. coli serotype O/K Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173033
Code : LD
Additional Details : Weight : 0.01000kg
Description
The PA1-73033 antibody reacts with E. coli. PA1-73033 has been successfully used in immunofluorescence applications. The PA1-73033 antibody was raised against E. coli serotypes O157:H7, O20, O125, O55, O111 & K12.
E. coli serotype O/K Polyclonal antibody specifically detects E. coli serotype O/K in Bacteria samples. It is validated for Immunocytochemistry.Specifications
E. coli serotype O/K | |
Polyclonal | |
FITC | |
Escherichia coli | |
E. coli serotypes O157:H7, O20, O125, O55, O111 & K12. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |