missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-cadherin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511549
Code : LE
Additional Details : Weight : 0.01000kg
Description
E-cadherin Polyclonal antibody specifically detects E-cadherin in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
E-cadherin | |
Polyclonal | |
Unconjugated | |
CDH1 | |
AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
999 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
1.6 mg/mL | |
PBS with 0.09% sodium azide | |
P12830 | |
CDH1 | |
KLH conjugated synthetic peptide between 833-862 amino acids from the C-terminal region of human E Cadherin (CDH1) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |