Learn More
Dynein Monoclonal Antibody (74.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1070
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-070 detects two isoforms of the ∽74 kDa polypetide of cytoplasmic dynein. MA1-070 has been successfully used in Western blot immunofluorescence, and immunoprecipitation procedures. In Western blot analysis of HeLa cell lysate this antibody detects a ∽74 kDa protein representing cytoplasmic dynein. The MA1-070 immunogen is purified bovine brain cytoplasmic dynein.
Dynein Monoclonal antibody specifically detects Dynein in Bovine, Human, Many, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot.Specifications
Dynein | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P61285, P63167, P63168, P63170, Q3MHR3, Q78P75, Q96FJ2, Q9D0M5 | |
DYNLL1, DYNLL2 | |
Purified bovine brain cytoplasmic dynein. | |
100 μg | |
Primary | |
Bovine, Human, Multi-species, Mouse, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot | |
74.1 | |
Unconjugated | |
DYNLL1 | |
1700064A15Rik; 6720463E02Rik; 8 kDa dynein light chain; 8 kDa dynein light chain b; 8kD LC; 8kDa LC; 8kDLC; C87222; CDLC2; cytoplasmic dynein light polypeptide; cytoplasmic dynein light polypeptide 1; cytoplasmic light-chain dynein; cytosolic DH IC-1; DLC1; Dlc2; DLC8; DLC8b; DNCL1; DNCL1B; Dnclc1; dynein LC8; dynein light chain 1, cytoplasmic; Dynein light chain 2, cytoplasmic; dynein light chain LC8-type 1; Dynein light chain LC8-type 2; dynein light chain-2; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; dynein, light chain, LC8-type 2; DYNLL1; DYNLL2; HDLC1; LC8; LC8; PIN; LC8a; LOC540369 protein; mPIN; PIN; Protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase; radial spoke 22 homolog; RSPH22; similar to cytoplasmic dynein light chain 1 | |
Mouse | |
Protein A | |
RUO | |
140734, 140735, 404151, 540369, 56455, 58945, 68097, 8655 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |