Learn More
Dynactin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530389
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: MCF-7. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (80%), Chimpanzee (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dynactin 2 Polyclonal antibody specifically detects Dynactin 2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Dynactin 2 | |
Polyclonal | |
Unconjugated | |
DCTN2 | |
2310042E05Rik; 50 kDa dynein-associated polypeptide; C130077D06Rik; C87049; DCTN2; dctn2 protein; DCTN50; DCTN-50; dynactin 2; dynactin 2 (p50); dynactin complex 50 kDa subunit; Dynactin subunit 2; dynamitin; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; HEL-S-77; hypothetical protein LOC527201; p50; p50 dynamitin; p50/dynamitin; RBP50; zgc:63867 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13561 | |
DCTN2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 253 of Human Dynactin 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |