Learn More
DUSP10 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA131512
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX22272-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DUSP10 Polyclonal antibody specifically detects DUSP10 in Human samples. It is validated for Western Blot.Specifications
DUSP10 | |
Polyclonal | |
Unconjugated | |
DUSP10 | |
2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase | |
Goat | |
IgG fraction | |
RUO | |
11221 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y6W6 | |
DUSP10 | |
Peptide with sequence RILTPKLMGVETVV, from C Terminus of the protein sequence according to NP_009138; NP_653329; NP_653330 | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |