Learn More
Dtk Rat anti-Mouse, Clone: 109646, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB759
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13431826
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Dtk Monoclonal antibody MAB759 Specifically detects Dtk in Mouse samples. It is validated for ELISA Capture, Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications
Specifications
Dtk/TYRO3
109646
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P55144
Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Tyro3/Dtk extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Tyro3/Dtk, recombinant mouse (rm) Mer, or rmAxl is observed.
Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Tyro3
Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk Ala31-Ser418 Accession # P55144
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7301
Reconstitute at 0.5 mg/mL in sterile PBS.