missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-DRP1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
POA
Specifications
Host Species | Rabbit |
---|---|
Isotype | IgG |
Immunogen | A synthetic peptide made to an internal region within residues 500-600 of the human DRP1 protein. [Swiss-Prot# O00429] |
Antigen | DRP1 |
Primary or Secondary | Primary |
Description
DRP1 Polyclonal specifically detects DRP1 in Human, Mouse, Rat, Fish, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Rabbit | |
A synthetic peptide made to an internal region within residues 500-600 of the human DRP1 protein. [Swiss-Prot# O00429] | |
Primary | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
O00429 | |
PBS with 0.02% Sodium Azide |
IgG | |
DRP1 | |
RUO | |
Human, primate, rat, mouse. Predicted to react with bovine based on 100% sequence homology. Fish reactivity reported in scientific literature (PMID: 25008790). | |
10059 | |
1 mg/mL |
For Research Use Only