missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPYSL4, Mouse, Clone: 1F5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DPYSL4.
Brand: Abnova H00010570-M01.100ug
Additional Details : Weight : 0.00010kg
Description
Sequence: VTPGAGRFVPRKTFPDFVYKRIKARNRLAEIHGVPRGLYDGPVHEVMVPAKPGSGAPARASCPGKISVPPVRNLHQSGFSLSGSQADDHIARRTAQKIMSpecifications
DPYSL4 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006426 | |
DPYSL4 | |
DPYSL4 (NP_006417, 461 a.a. ∼ 559 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunoprecipitation, Western Blot | |
1F5 | |
Mouse monoclonal antibody raised against a partial recombinant DPYSL4. | |
DPYSL4 | |
CRMP3/DRP-4/ULIP4 | |
Mouse | |
Affinity chromatography | |
RUO | |
10570 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |