Learn More
DPPIV/CD26 Rat anti-Mouse, Clone: 155211, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB954
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13462036
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
DPPIV/CD26 Monoclonal antibody MAB954 Specifically detects DPPIV/CD26 in Mouse samples. It is validated for ELISA Capture, Immunoprecipitation, Western Blotting
Specifications
Specifications
DPPIV/CD26
155211
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P28843
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse DPPIV/CD26 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse Cathepsin A or recombinant human DPPIV is observed.
Mouse
Western Blot, Immunoprecipitation, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DPP4
Mouse myeloma cell line NS0-derived recombinant mouse DPPIV/CD26 Ser29-His760 (predicted) Accession # P28843
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1803
Reconstitute at 0.5 mg/mL in sterile PBS.