missing translation for 'onlineSavingsMsg'
Learn More

DPPII/QPP/DPP7 Rat anti-Human, Clone: 398024, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB34381

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13430076

  • £286.65 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DPPII/QPP/DPP7 Monoclonal antibody MAB34381 Specifically detects DPPII/QPP/DPP7 in Human samples. It is validated for Western Blotting
Specifications

Specifications

DPPII/QPP/DPP7
398024
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DPP7
Mouse myeloma cell line NS0-derived recombinant human DPPII/QPP/DPP7 Gly22-Leu492 Accession # AAH11907
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
29952
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAH11907
carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPP7, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human DPPII/QPP/DPP7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 5% cross-reactivity with recombinant mouse DPP7 is observed.
Human
Videos
Documents

For Research Use Only