Learn More
DPPA5/ESG1 Mouse anti-Human, Clone: 319606, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3125
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439905
  • £288.60 / 100µg
Estimated Shipment
18-08-2020
Description
Description
DPPA5/ESG1 Monoclonal antibody MAB3125 Specifically detects DPPA5/ESG1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
DPPA5/ESG1
319606
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DPPA5
E. coli-derived recombinant human DPPA5/ESG1 Met1-Lys116 Accession # A6NC42
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
340168
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
A6NC42
developmental pluripotency associated 5, developmental pluripotency-associated 5 protein, embryonal stem cell specific gene 1, Embryonal stem cell-specific gene 1 protein, Esg1, ESG1ESG-1, hDPPA5
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human DPPA5/ESG1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human DPPA4 or recombinant mouse DPPA5.