Learn More
DPP6 Mouse anti-Human, Clone: 274311, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB23601
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439385
  • £288.60 / 100µg
Estimated Shipment
18-08-2020
Description
Description
DPP6 Monoclonal antibody MAB23601 Specifically detects DPP6 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
DPP6
274311
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DPP6
Mouse myeloma cell line NS0-derived recombinant human DPP6 Thr118-Asp865 Accession # P42658
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1804
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P42658
dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPX, DPPXFLJ55680, MGC46605, VF2
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human DPP6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human DPPIV/CD26 is observed.